COM448671572 Combi-Blocks, Inc.

15-Bromo-1-pentadecanol

  • MDL number MFCD00482036
  • Cas Number : 59101-27-8
  • Molecular Weight : 307.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-1766

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-1766
Availability details for
Quantity
  • Availability
  • Add to Cart