COM448671641 Combi-Blocks, Inc.

1-Benzyl-2-methylpiperidin-4-one, HCl

  • MDL number MFCD19382499
  • Cas Number : 203661-73-8
  • Molecular Weight : 239.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-1852

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-1852
Availability details for
Quantity
  • Availability
  • Add to Cart