COM448671661 Combi-Blocks, Inc.

1-Benzyl-4-(pyridin-2-yl)piperidin-4-ol

  • MDL number MFCD09788612
  • Cas Number : 65869-51-4
  • Molecular Weight : 268.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-1881

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-1881
Availability details for
Quantity
  • Availability
  • Add to Cart