COM448671664 Combi-Blocks, Inc.

1-Benzyl-4-boc-piperazine-2-carboxylic acid

  • MDL number MFCD09800391
  • Cas Number : 181956-25-2
  • Molecular Weight : 320.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-1886

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-1886
Availability details for
Quantity
  • Availability
  • Add to Cart