COM448671725 Combi-Blocks, Inc.

1-Boc-3-n-(pyridin-4-ylmethyl)-amino-pyrrolidine

  • MDL number MFCD07369887
  • Cas Number : 886364-95-0
  • Molecular Weight : 277.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-1971

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-1971
Availability details for
Quantity
  • Availability
  • Add to Cart