COM448671748 Combi-Blocks, Inc.

1-Boc-4-(4-bromo-butyl)-piperidine

  • MDL number MFCD07369864
  • Cas Number : 142355-81-5
  • Molecular Weight : 320.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2003

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2003
Availability details for
Quantity
  • Availability
  • Add to Cart