COM448671754 Combi-Blocks, Inc.

1-Boc-4-methylazepane-4-carboxylic acid

  • MDL number MFCD10687157
  • Cas Number : 1027512-23-7
  • Molecular Weight : 257.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2011

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2011
Availability details for
Quantity
  • Availability
  • Add to Cart