COM448671761 Combi-Blocks, Inc.

1-Boc-6-amino-3,3-dimethyl-2-oxo-2,3-dihydro-indole

  • MDL number MFCD11053532
  • Cas Number : 1049677-44-2
  • Molecular Weight : 276.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2022

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2022
Availability details for
Quantity
  • Availability
  • Add to Cart