COM448671778 Combi-Blocks, Inc.

1-Bromo-3,3-diphenylpropane

  • MDL number MFCD00044829
  • Cas Number : 20017-68-9
  • Molecular Weight : 275.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2041

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2041
Availability details for
Quantity
  • Availability
  • Add to Cart