COM448671842 Combi-Blocks, Inc.

1-Cbz-piperidin-4-ylidene-acetic acid

  • MDL number MFCD04115020
  • Cas Number : 40113-03-9
  • Molecular Weight : 275.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2123

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2123
Availability details for
Quantity
  • Availability
  • Add to Cart