COM448671904 Combi-Blocks, Inc.

1-Ethyloxycarbonyl-3-[4-(4-methyl-piperazin-1-yl)-benzoylamino]-5-boc-4,6-dihydro-pyrrolo[3,4-c]pyrazole

  • MDL number MFCD11053614
  • Cas Number : 761443-69-0
  • Molecular Weight : 498.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2201

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2201
Availability details for
Quantity
  • Availability
  • Add to Cart