COM448671990 Combi-Blocks, Inc.

1-Isobutyl-l-proline

  • MDL number MFCD11053658
  • Cas Number : 1044637-62-8
  • Molecular Weight : 171.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2308

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2308
Availability details for
Quantity
  • Availability
  • Add to Cart