COM448672034 Combi-Blocks, Inc.

1-Methyl-3-hydroxy-2(1H)-pyridinone

  • MDL number MFCD11848440
  • Cas Number : 19365-01-6
  • Molecular Weight : 125.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2360

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2360
Availability details for
Quantity
  • Availability
  • Add to Cart