COM448672074 Combi-Blocks, Inc.

1-Methylazepane-4-carboxylic acid

  • MDL number MFCD10687192
  • Cas Number : 1027512-62-4
  • Molecular Weight : 157.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2408

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2408
Availability details for
Quantity
  • Availability
  • Add to Cart