COM448672084 Combi-Blocks, Inc.

1-Naphthol-4-sulfonic acid

  • MDL number MFCD00065324
  • Cas Number : 84-87-7
  • Molecular Weight : 224.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2420

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2420
Availability details for
Quantity
  • Availability
  • Add to Cart