COM448672136 Combi-Blocks, Inc.

1-N-Boc-4-(4-carbamoylpyridin-2-yl)piperazine

  • MDL number MFCD12068360
  • Cas Number : 1159823-86-5
  • Molecular Weight : 306.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2481

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2481
Availability details for
Quantity
  • Availability
  • Add to Cart