COM448672139 Combi-Blocks, Inc.

1-N-Boc-4-(4-chlorophenyl)piperidine

  • MDL number MFCD09878786
  • Cas Number : 946593-11-9
  • Molecular Weight : 295.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2484

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2484
Availability details for
Quantity
  • Availability
  • Add to Cart