COM448672169 Combi-Blocks, Inc.

1-N-Fmoc-3-pyrrolidinone

  • MDL number MFCD01861826
  • Cas Number : 672310-12-2
  • Molecular Weight : 307.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2518

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2518
Availability details for
Quantity
  • Availability
  • Add to Cart