COM448672182 Combi-Blocks, Inc.

1-Phenyl-1,3,8-triazaspiro[4.5]decan-4-one

  • MDL number MFCD00005977
  • Cas Number : 1021-25-6
  • Molecular Weight : 231.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2533

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2533
Availability details for
Quantity
  • Availability
  • Add to Cart