COM448672185 Combi-Blocks, Inc.

1-Phenyl-1h-imidazol-4-amine, HCl

  • MDL number MFCD20921642
  • Cas Number : 158688-63-2
  • Molecular Weight : 195.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2537

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2537
Availability details for
Quantity
  • Availability
  • Add to Cart