COM448672206 Combi-Blocks, Inc.

1-Phenyl-3-(1-(2,2,2-trifluoroacetyl)pyrrolidin-3-yl)urea

  • MDL number MFCD09788592
  • Cas Number : 951623-94-2
  • Molecular Weight : 301.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2561

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2561
Availability details for
Quantity
  • Availability
  • Add to Cart