COM448672252 Combi-Blocks, Inc.

1-Pyrrolidin-2-(n-cbz-n-methyl)amino-ethane

  • MDL number MFCD03839945
  • Cas Number : 886362-96-5
  • Molecular Weight : 262.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2614

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2614
Availability details for
Quantity
  • Availability
  • Add to Cart