COM448672270 Combi-Blocks, Inc.

1-tert-Butyl 2-ethyl 2,4,4-trimethyl-5-oxopyrrolidine-1,2-dicarboxylate

  • MDL number MFCD18711598
  • Cas Number : 1001353-88-3
  • Molecular Weight : 299.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2633

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2633
Availability details for
Quantity
  • Availability
  • Add to Cart