COM448672274 Combi-Blocks, Inc.

1-tert-Butyl 2-methyl 2-methylpiperidine-1,2-dicarboxylate

  • MDL number MFCD18642716
  • Cas Number : 470668-97-4
  • Molecular Weight : 257.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2637

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2637
Availability details for
Quantity
  • Availability
  • Add to Cart