COM448672275 Combi-Blocks, Inc.

1-tert-Butyl 2-methyl azepane-1,2-dicarboxylate

  • MDL number MFCD20921681
  • Cas Number : 1352305-12-4
  • Molecular Weight : 257.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2638

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2638
Availability details for
Quantity
  • Availability
  • Add to Cart