COM448672340 Combi-Blocks, Inc.

2-(1-Methyl-1h-pyrazol-3-yl)phenol

  • MDL number MFCD19382470
  • Cas Number : 123532-18-3
  • Molecular Weight : 174.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2723

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2723
Availability details for
Quantity
  • Availability
  • Add to Cart