COM448672352 Combi-Blocks, Inc.

2-(2-(Pyridin-3-yl)pyrrolidin-2-yl)ethanamine tri, HCl

  • MDL number MFCD20921687
  • Cas Number : 1352305-25-9
  • Molecular Weight : 300.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2738

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2738
Availability details for
Quantity
  • Availability
  • Add to Cart