COM448672362 Combi-Blocks, Inc.

2-(2,4,5-Trifluorophenyl)-ethanol

  • MDL number MFCD06660356
  • Cas Number : 883267-70-7
  • Molecular Weight : 176.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2748

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2748
Availability details for
Quantity
  • Availability
  • Add to Cart