COM448672395 Combi-Blocks, Inc.

2-(2-Chloro-ethyl)-5-fluoro-isoindole-1,3-dione

  • MDL number MFCD06796531
  • Cas Number : 176200-91-2
  • Molecular Weight : 227.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2786

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2786
Availability details for
Quantity
  • Availability
  • Add to Cart