COM448672465 Combi-Blocks, Inc.

2-(2-Cyanophenoxy)-4-chloropyrimidine

  • MDL number MFCD12068369
  • Cas Number : 1159826-56-8
  • Molecular Weight : 231.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2862

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2862
Availability details for
Quantity
  • Availability
  • Add to Cart