COM448672473 Combi-Blocks, Inc.

2-(2-Isopropylamino-thiazol-4-yl)-7-methoxy-quinolin-4-ol

  • MDL number MFCD04974520
  • Cas Number : 300831-05-4
  • Molecular Weight : 315.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2873

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2873
Availability details for
Quantity
  • Availability
  • Add to Cart