COM448672474 Combi-Blocks, Inc.

2-(2-Methoxybenzylamino)nicotinic acid

  • MDL number MFCD11040502
  • Cas Number : 1019372-81-6
  • Molecular Weight : 258.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2874

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2874
Availability details for
Quantity
  • Availability
  • Add to Cart