COM448672488 Combi-Blocks, Inc.

2-(2-Methylphenoxy)benzylamine, HCl

  • MDL number MFCD03840024
  • Cas Number : 870061-76-0
  • Molecular Weight : 249.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2893

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2893
Availability details for
Quantity
  • Availability
  • Add to Cart