COM448672498 Combi-Blocks, Inc.

2-(2-Thienyl)-2-[4-(trifluoromethyl)phenyl]acetic acid

  • MDL number MFCD01631969
  • Cas Number : 885950-82-3
  • Molecular Weight : 286.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2904

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2904
Availability details for
Quantity
  • Availability
  • Add to Cart