COM448672506 Combi-Blocks, Inc.

2-(3-(2-Chlorophenyl)ureido)propanoic acid

  • MDL number MFCD09998256
  • Cas Number : 1009544-03-9
  • Molecular Weight : 242.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2913

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2913
Availability details for
Quantity
  • Availability
  • Add to Cart