COM448672512 Combi-Blocks, Inc.

2-(3,4-Dichlorophenyl)-2-phenylacetonitrile

  • MDL number MFCD09788610
  • Cas Number : 92150-76-0
  • Molecular Weight : 262.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2920

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2920
Availability details for
Quantity
  • Availability
  • Add to Cart