COM448672514 Combi-Blocks, Inc.

2-(3,4-Dichlorophenyl)-2-phenylethylamine, HCl

  • MDL number MFCD02089462
  • Cas Number : 1021871-57-7
  • Molecular Weight : 302.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2922

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2922
Availability details for
Quantity
  • Availability
  • Add to Cart