COM448672515 Combi-Blocks, Inc.

2-(3,4-Dichloro-phenyl)-oxirane

  • MDL number MFCD07784084
  • Cas Number : 52909-94-1
  • Molecular Weight : 189.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2923

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-2923
Availability details for
Quantity
  • Availability
  • Add to Cart