COM448672604 Combi-Blocks, Inc.

2-(3-Methoxy-4-bromophenyl)-ethylamine hcl

  • MDL number MFCD11053673
  • Cas Number : 1159826-47-7
  • Molecular Weight : 266.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-3036

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-3036
Availability details for
Quantity
  • Availability
  • Add to Cart