COM448672605 Combi-Blocks, Inc.

2-(3-Methylbut-2-enyloxy)isoindoline-1,3-dione

  • MDL number MFCD11100906
  • Cas Number : 157128-91-1
  • Molecular Weight : 231.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-3040

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-3040
Availability details for
Quantity
  • Availability
  • Add to Cart