COM448672615 Combi-Blocks, Inc.

2-(4-1,2,3-Thiadiazolyl)pyridine

  • MDL number MFCD01109388
  • Cas Number : 176037-42-6
  • Molecular Weight : 163.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-3059

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-3059
Availability details for
Quantity
  • Availability
  • Add to Cart