COM448672619 Combi-Blocks, Inc.

2-(4-Boc-piperazinyl)-4-phenylbutanoic acid

  • MDL number MFCD01862502
  • Cas Number : 885274-45-3
  • Molecular Weight : 348.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-3065

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-3065
Availability details for
Quantity
  • Availability
  • Add to Cart