COM448672627 Combi-Blocks, Inc.

2-(4-Chlorophenyl)-2-(4-methylpiperazin-1-yl)acetamide

  • MDL number MFCD09998242
  • Cas Number : 1015856-12-8
  • Molecular Weight : 267.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-3080

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-3080
Availability details for
Quantity
  • Availability
  • Add to Cart