COM448672644 Combi-Blocks, Inc.

2-(4-Fluorophenoxy)-1-propanamine, HCl

  • MDL number MFCD03002051
  • Cas Number : 1051368-76-3
  • Molecular Weight : 205.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-3108

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-3108
Availability details for
Quantity
  • Availability
  • Add to Cart