COM448672824 Combi-Blocks, Inc.

2-(4-Benzyloxy-phenyl)-ethylamine, HCl

  • MDL number MFCD01657171
  • Cas Number : 2982-54-9
  • Molecular Weight : 263.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-3292

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-3292
Availability details for
Quantity
  • Availability
  • Add to Cart