COM448672851 Combi-Blocks, Inc.

2-(4-Fluorophenyl)-2-phenyl-acetonitrile

  • MDL number MFCD00019794
  • Cas Number : 719-82-4
  • Molecular Weight : 211.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-3319

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-3319
Availability details for
Quantity
  • Availability
  • Add to Cart