COM448672947 Combi-Blocks, Inc.

2-Imidazol-1-yl-benzaldehyde

  • MDL number MFCD05864671
  • Cas Number : 151055-86-6
  • Molecular Weight : 172.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-3421

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-3421
Availability details for
Quantity
  • Availability
  • Add to Cart