COM448673008 Combi-Blocks, Inc.

2-(4-Fluorophenyl)thiomorpholine

  • MDL number MFCD03002056
  • Cas Number : 1001940-38-0
  • Molecular Weight : 197.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-3484

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-3484
Availability details for
Quantity
  • Availability
  • Add to Cart