COM448673011 Combi-Blocks, Inc.

2-(4-Hydroxy-phenyl)-n-phenyl-acetamide

  • MDL number MFCD06796563
  • Cas Number : 131179-71-0
  • Molecular Weight : 227.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-3487

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-3487
Availability details for
Quantity
  • Availability
  • Add to Cart