COM448673013 Combi-Blocks, Inc.

2-(4-Methoxyphenoxy)benzylamine, HCl

  • MDL number MFCD03840019
  • Cas Number : 1171721-42-8
  • Molecular Weight : 265.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-3489

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-3489
Availability details for
Quantity
  • Availability
  • Add to Cart